Learn More
Barium nitrate, 99.95% (metals basis), Thermo Scientific Chemicals
£34.57 - £112.56
Chemical Identifiers
CAS | 10022-31-8 |
---|---|
Molecular Formula | BaN2O6 |
Molecular Weight (g/mol) | 261.34 |
MDL Number | MFCD00003442 |
InChI Key | IWOUKMZUPDVPGQ-UHFFFAOYSA-N |
Synonym | barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 |
PubChem CID | 24798 |
IUPAC Name | barium(2+) dinitrate |
SMILES | [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367278
|
Thermo Scientific Alfa Aesar
030481.14 |
25 g |
£34.57
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377278
|
Thermo Scientific Alfa Aesar
030481.22 |
100 g |
£112.56
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium nitrate is used in the manufacture of special glasses, in fire works industries and for green-fire in pyrotechnics. It is widely used in defense applications. It is a precursor material for barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10022-31-8 | |
261.34 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
24798 | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
BaN2O6 | |
MFCD00003442 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
barium(2+) dinitrate |
Specifications
592°C (decomposition) | |
White | |
99.95% | |
MFCD00003442 | |
Soluble in water. Insoluble in alcohol. | |
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
261.34 | |
261.35 | |
White crystalline | |
Barium nitrate |
10022-31-8 | |
Crystalline | |
BaN2O6 | |
barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2 | |
IWOUKMZUPDVPGQ-UHFFFAOYSA-N | |
barium(2+) dinitrate | |
24798 | |
Hygroscopic | |
3.24 g/cm3 |
Safety and Handling
P210-P220-P221-P261-P264b-P270-P271-P280-P301+P310-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
H272-H301-H319-H332
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II
missing translation for 'einecsNumber' : 233-020-5
missing translation for 'rtecsNumber' : CQ9625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only