missing translation for 'onlineSavingsMsg'
Learn More
Barium nitrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10369183

Barium nitrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10369183
2.5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10369183

Brand: Thermo Scientific Acros 203150025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Synonym barium nitrate, nitrobarite, nitric acid, barium salt, barium dinitrate, bariumnitrate, dusicnan barnaty czech, nitrato barico spanish, unii-mdc5sw56xc, nitrate de baryum french, barium nitrate ba no3 2
PubChem CID 24798
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592.0°C
Color White
pH 5 to 8 (5% soln. at 20°C)
Physical Form Crystals
Quantity 2.5 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula Ba(NO3)2
Merck Index 15, 979
Solubility Information Solubility in water: 9g/100mL (20°C). Other solubilities: practically insoluble in alcohol and acetone
IUPAC Name barium(2+);dinitrate
Formula Weight 261.34
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 3.23
Chemical Name or Material Barium nitrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464852 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number203155000,203150025
Lot NumberA0458912 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog Number203155000,203150025
Lot NumberA0413330 Certificate TypeCertificate of Analysis Date25/04/2022 Catalog Number203155000,203150025
Lot NumberA0437332 Certificate TypeCertificate of Analysis Date17/12/2021 Catalog Number203155000,203150025
Lot NumberA0394833 Certificate TypeCertificate of Analysis Date14/09/2020 Catalog Number
compliance-icons compliance-icons
Product Identifier
  • Barium nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium nitrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.