missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Nitrate, ACS Reagent, ≥99%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 592°C |
Color | Colorless |
Quantity | 100 g |
Linear Formula | Ba(NO3)2 |
UN Number | UN1446 |
Formula Weight | 261.34g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Appearance | Crystals |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'