missing translation for 'onlineSavingsMsg'
Learn More

Barium Nitrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10590521
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10022-31-8
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
IUPAC Name barium(2+) dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 592°C
Color White
pH 5.0 to 8.0
Physical Form Solid
Quantity 1 kg
Formula Weight 261.34g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Barium nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'