missing translation for 'onlineSavingsMsg'
Learn More

Barium oxide, 99.5% (metals basis excluding Sr), Sr <500ppm

Product Code. 11450740
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11450740

Brand: Thermo Scientific Alfa Aesar 044440.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1304-28-5 | BaO | 153.33 g/mol

Barium oxide is used in cathode ray tubes, crown glass, and catalysts. It is used for the adsorption NO and NO2. It also plays an important role for NO2 storage and reduction. It acts as an efficient ethoxylation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-28-5
Molecular Formula BaO
Molecular Weight (g/mol) 153.33
MDL Number MFCD00003453
InChI Key QVQLCTNNEUAWMS-UHFFFAOYSA-N
Synonym barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum
PubChem CID 62392
IUPAC Name oxobarium
SMILES O=[Ba]

Specifications

Melting Point 1923°C
Physical Form Powder
Quantity 100 g
UN Number UN1884
Merck Index 14,986
Solubility Information Sparingly soluble in water. Soluble in ethanol,dilute mineral acids and alkali.
Formula Weight 153.33
Percent Purity 99.5%
Refractive Index 1.98
Density 5.7 g/mL
Assay (metals basis excluding Sr), Sr <500ppm
Chemical Name or Material Barium oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Barium oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Barium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.