Learn More
Barium oxide, 90%, extra pure, powder, Thermo Scientific Chemicals
£25.20 - £229.00
Chemical Identifiers
CAS | 1304-28-5, 513-77-9, 1304-29-6 |
---|---|
Molecular Formula | BaO |
Molecular Weight (g/mol) | 153.33 |
MDL Number | MFCD00003453 |
InChI Key | QVQLCTNNEUAWMS-UHFFFAOYSA-N |
Synonym | barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum |
PubChem CID | 62392 |
IUPAC Name | oxobarium |
SMILES | O=[Ba] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10424555
|
Thermo Scientific Acros
207830250 |
25 g | Glass Bottle | ||||||
10104600
|
Thermo Scientific Acros
207835000 |
500 g | Plastic bottle | ||||||
11994131
|
Thermo Scientific Acros
207830025 |
2.5 kg | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1304-28-5, 513-77-9, 1304-29-6 | |
153.33 | |
QVQLCTNNEUAWMS-UHFFFAOYSA-N | |
62392 | |
O=[Ba] |
BaO | |
MFCD00003453 | |
barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum | |
oxobarium |
Specifications
2000.0°C | |
1304-28-5, 513-77-9, 1304-29-6 | |
Powder | |
BaO | |
MFCD00003453 | |
15,982 | |
Solubility in water: soluble. Other solubilities: soluble in diluted acids, alcohols, insoluble in acetone, nh3, slowly but considerably soluble in methanol,, ethanol forming barium alcoholate | |
O=[Ba] | |
153.33 | |
153.34 | |
Extra Pure |
1920.0°C | |
Gray to Green-Yellow | |
90% | |
BaO | |
01,683; 09,23 | |
barium oxide, baryta, barium monoxide, calcined baryta, barium protoxide, oxyde de baryum french, barium oxide, obtained by calcining witherite, oxyde de baryum | |
QVQLCTNNEUAWMS-UHFFFAOYSA-N | |
oxobarium | |
62392 | |
90% | |
Barium oxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 215-127-9
- Barium oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : Barium oxide, bariumcarbonate
RUO – Research Use Only