missing translation for 'onlineSavingsMsg'
Learn More
Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals Product Code.: 10730731

Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10730731
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10730731

Brand: Thermo Scientific Acros 221275000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-95-7
Molecular Formula BaCl2O8
Molecular Weight (g/mol) 336.23
MDL Number MFCD00003440
InChI Key OOULUYZFLXDWDQ-UHFFFAOYSA-L
Synonym barium perchlorate, barium diperchlorate, perchloric acid, barium salt, unii-ey48pa0c98, barium 2+ diperchlorate, barium 2+ ion diperchlorate, barium perchlorate, anhydrous, perchloric acid, bariumsalt 2:1, perchloric acid, barium salt 2:1, acmc-20alqv
PubChem CID 61623
IUPAC Name barium(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ba+2]

Specifications

Melting Point 505.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ba(ClO4)2
Merck Index 15, 983
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, slightly soluble in ethanol, ethyl acetate and, acetone
Formula Weight 336.23
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 3.2
Density 3.2000g/mL
Chemical Name or Material Barium perchlorate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461866 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog Number221275000,221271000
Lot NumberA0448462 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number221275000,221271000
Lot NumberA0457328 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number221275000,221271000
Lot NumberA0403984 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number221275000,221271000
Lot NumberA0412766 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number221275000,221271000
compliance-icons compliance-icons
Product Identifier
  • Barium perchlorate, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium perchlorate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.