missing translation for 'onlineSavingsMsg'
Learn More

Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%

Product Code. 11366607
Click to view available options
Quantity:
5 g
50 g
250 g
Unit Size:
250g
50g
5g
This item is not returnable. View return policy

Product Code. 11366607

Brand: Thermo Scientific Alfa Aesar 000653.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-39-3 | Ba | 137.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atom
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Quantity 250 g
Odor Odorless
Assay Percent Range 99.2% (Metals basis)
Form Pieces
Chemical Name or Material Barium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.