Learn More
Barium silicon oxide, Thermo Scientific Chemicals
£27.74 - £163.82
Chemical Identifiers
CAS | 13255-26-0 |
---|---|
Molecular Formula | BaO3Si |
Molecular Weight (g/mol) | 213.409 |
MDL Number | MFCD00014186 |
InChI Key | HMOQPOVBDRFNIU-UHFFFAOYSA-N |
Synonym | barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1Show More |
PubChem CID | 166725 |
IUPAC Name | barium(2+);dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[Ba+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11367098
|
Thermo Scientific Alfa Aesar
015122.22 |
100 g | N/A | N/A | N/A | |||||
11357098
|
Thermo Scientific Alfa Aesar
015122.36 |
500 g |
£163.82
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Barium silicate films that are suitable for use as transmission media in integrated optical circuits. It is shown that the films, which can be produced with a wide range of refractive indices by suitable selection of the ratio of the target constituents, exhibit low optical attenuation. Hence it is used for integrated optical circuits. It is used in ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13255-26-0 | |
213.409 | |
HMOQPOVBDRFNIU-UHFFFAOYSA-N | |
166725 | |
[O-][Si](=O)[O-].[Ba+2] |
BaO3Si | |
MFCD00014186 | |
barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1Show More | |
barium(2+);dioxido(oxo)silane |
Specifications
13255-26-0 | |
Powder | |
MFCD00014186 | |
HMOQPOVBDRFNIU-UHFFFAOYSA-N | |
barium(2+);dioxido(oxo)silane | |
166725 | |
Barium silicon oxide |
White | |
BaO3Si | |
barium silicate, barium metasilicate, silicic acid, barium salt, barium silicon oxide, barium mesosilicate, barium disilicate, bariumsiliconoxide, ba.so3i, barium meta silicate, silicic acid h2sio3 , barium salt 1:1 | |
[O-][Si](=O)[O-].[Ba+2] | |
213.409 | |
213.4 |
Safety and Handling
H302-H332: Harmful if swallowed. Harmful if inhaled.
missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: III
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.