missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 500 mL |
Color | Colorless |
pH | 1 |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'