missing translation for 'onlineSavingsMsg'
Learn More
Barium sulfate, 97%, Thermo Scientific Chemicals Product Code.: 11381499

Barium sulfate, 97%, Thermo Scientific Chemicals

Product Code. 11381499
100 g
Click to view available options
Quantity:
100 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11381499

Brand: Thermo Scientific Alfa Aesar 013989.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, and as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C (decomposition)
Physical Form Powder
Quantity 100 g
Solubility Information Insoluble in water,dilute acids,alcohol. Soluble in hot,concentrated H2SO4
IUPAC Name barium(2+) sulfate
Formula Weight 233.4
Density 4.5
Chemical Name or Material Barium sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN25L008 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog Number013989.22,013989.A1,013989.A7
Lot NumberS28K037 Certificate TypeCertificate of Analysis Date25/07/2024 Catalog Number013989.22,013989.A1,013989.A7
Lot NumberU28J018 Certificate TypeCertificate of Analysis Date15/09/2023 Catalog Number013989.22,013989.A1,013989.A7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium sulfate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.