missing translation for 'onlineSavingsMsg'
Learn More
Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™ Product Code.: 15621050

Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™
BIOPHARMA

Product Code. 15621050 Shop All Honeywell Fluka Products
1 kg
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
This item is not returnable. View return policy

Product Code. 15621050

Brand: Honeywell-Fluka 114321KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., 97.5-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
IUPAC Name barium(2+);sulfate
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula BaSO4
UN Number NONH for all modes of transport
Formula Weight 233.39g/mol
Percent Purity 97.5 to 100.5%
Grade puriss.
Chemical Name or Material Barium Sulfate, Meets analytical specification of Ph.Eur.BP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.