missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium sulfate, Puriss., meets analytical spec. of BP, Ph. Eur., 97.5-100.5%, Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Max % | 100.0000% |
Quantity | 25 kg |
Assay Percent Range | 97.5 to 100.5% |
Linear Formula | BaSO4 |
UN Number | NONH for all modes of transport |
Formula Weight | 233.39 |
Percent Purity | 97.5 to 100.5% |
Grade | puriss. |
Packaging | Paper / Poly Bag |
Density | 4.400 g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'