missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Sulfate Precipitated, Extra Pure, SLR, Fisher Chemical™
£54.60 - £107.12
Chemical Identifiers
CAS | 7727-43-7 |
---|---|
Molecular Formula | BaO4S |
Molecular Weight (g/mol) | 233.38 |
MDL Number | MFCD00003455 |
InChI Key | TZCXTZWJZNENPQ-UHFFFAOYSA-L |
IUPAC Name | barium(2+) sulfate |
SMILES | [Ba++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10365550
|
Fisher Chemical
B/1120/53 |
500 g | N/A | ||||||
12618625
|
Fisher Chemical
B/1120/61 |
2 kg |
£107.12
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
7727-43-7 | |
233.38 | |
TZCXTZWJZNENPQ-UHFFFAOYSA-L | |
[Ba++].[O-]S([O-])(=O)=O |
BaO4S | |
MFCD00003455 | |
barium(2+) sulfate |
Specifications
1580°C | |
White | |
BaO4S | |
TZCXTZWJZNENPQ-UHFFFAOYSA-L | |
barium(2+) sulfate | |
233.4g/mol |
7727-43-7 | |
Powder/Solid | |
MFCD00003455 | |
[Ba++].[O-]S([O-])(=O)=O | |
233.38 | |
Plastic powder jar |
Safety and Handling
Product Identifier
- Barium sulfate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Barium Sulfate Precipitated, Extra Pure, SLR, Fisher Chemical™