missing translation for 'onlineSavingsMsg'
Learn More

Barium sulfide, 99.7% (metals basis), Thermo Scientific Chemicals

£80.10 - £323.61

Chemical Identifiers

CAS 21109-95-5
Molecular Formula BaS
Molecular Weight (g/mol) 169.39
MDL Number MFCD00014192
InChI Key CJDPJFRMHVXWPT-UHFFFAOYSA-N
Synonym barium sulfide, thioxobarium
PubChem CID 88791
IUPAC Name barium(2+) sulfanediide
SMILES [S--].[Ba++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397577
SDS View Documents
Thermo Scientific Alfa Aesar
012848.09
10 g
£80.10
10g
Please to purchase this item. Need a web account? Register with us today!
11307587
SDS View Documents
Thermo Scientific Alfa Aesar
012848.18
50 g
£323.61
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium sulfide has different characters, so it is used in a variety of ways, such as in electronics, paint pigments, dehairing hides, flame retardant, luminous paints, and generating pure hydrogen sulfide. Barium sulfide can deviate X-rays and helps to give clearly image of the soft tissue. Barium sulfide is a precursor to other barium compounds and also used as a short wavelength emitters for electronic displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 244-214-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium sulfide, 99.7% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.