missing translation for 'onlineSavingsMsg'
Learn More
Barium, Technical, Metal In Liquid PARaffin, SLR, Fisher Chemical Product Code.: 10417250

Barium, Technical, Metal In Liquid PARaffin, SLR, Fisher Chemical

Product Code. 10417250
25g, Plastic container
Click to view available options
25g, Plastic container
This item is not returnable. View return policy

Product Code. 10417250

Brand: Fisher Chemical B/0150/46

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Gray
CAS 7440-39-3
Melting Point 725°C
Boiling Point 1640°C
Molecular Formula Ba
MDL Number MFCD00134031
Quantity 25 g
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
SMILES [Ba]
IUPAC Name barium
Molecular Weight (g/mol) 137.33
Formula Weight 137.34g/mol
Packaging Plastic container
Vapor Pressure 10mmHg 1094
Form Solid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Barium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H228-Flammable solid.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H301-Toxic if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use to extinguish.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium, Technical, Metal In Liquid PARaffin, SLR, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.