missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium titanium oxide, 99.95% (metals basis), Thermo Scientific Chemicals
£52.76 - £211.05
Chemical Identifiers
CAS | 12047-27-7 |
---|---|
Molecular Formula | BaO3Ti |
Molecular Weight (g/mol) | 233.19 |
MDL Number | MFCD00003447 |
InChI Key | WNKMTAQXMLAYHX-UHFFFAOYSA-N |
Synonym | Barium titanate |
IUPAC Name | barium(2+) oxotitaniumbis(olate) |
SMILES | [Ba++].[O-][Ti]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492137
|
Thermo Scientific Alfa Aesar
045083.06 |
5 g |
£52.76
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15402147
|
Thermo Scientific Alfa Aesar
045083.14 |
25 g |
£211.05
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12047-27-7 | |
233.19 | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) |
BaO3Ti | |
MFCD00003447 | |
Barium titanate | |
[Ba++].[O-][Ti]([O-])=O |
Specifications
1620°C | |
Pieces | |
MFCD00003447 | |
Barium titanate | |
[Ba++].[O-][Ti]([O-])=O | |
233.19 | |
99.95% | |
(metals basis) |
12047-27-7 | |
BaO3Ti | |
14,1000 | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) | |
233.19 | |
Odorless | |
Barium titanium oxide |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'einecsNumber' : 234-975-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only