Learn More
Barium titanium oxide, high surface area, Thermo Scientific Chemicals
£65.12 - £193.97
Chemical Identifiers
CAS | 12047-27-7 |
---|---|
Molecular Formula | BaO3Ti |
Molecular Weight (g/mol) | 233.19 |
MDL Number | MFCD00003447 |
InChI Key | WNKMTAQXMLAYHX-UHFFFAOYSA-N |
Synonym | Barium titanate |
IUPAC Name | barium(2+) oxotitaniumbis(olate) |
SMILES | [Ba++].[O-][Ti]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358768
|
Thermo Scientific Alfa Aesar
041633.14 |
25 g |
£65.12
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368768
|
Thermo Scientific Alfa Aesar
041633.22 |
100 g |
£193.97
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12047-27-7 | |
233.19 | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) |
BaO3Ti | |
MFCD00003447 | |
Barium titanate | |
[Ba++].[O-][Ti]([O-])=O |
Specifications
1620°C | |
White to Gray | |
BaO3Ti | |
14,1000 | |
Slightly soluble in dilute mineral acids. Soluble in concentrated hydrofluoric acid. Insoluble in water. | |
[Ba++].[O-][Ti]([O-])=O | |
233.19 | |
≥99% | |
5.85 g/mL | |
Barium titanium oxide, High surface area |
12047-27-7 | |
Powder | |
MFCD00003447 | |
Barium titanate | |
WNKMTAQXMLAYHX-UHFFFAOYSA-N | |
barium(2+) oxotitaniumbis(olate) | |
233.19 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'einecsNumber' : 234-975-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only