Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Brand:  Thermo Scientific™ D5028
Code : A0
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 11786259
  • £602.34 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
Specifications
Cartridge kit for E-Pure 4-holder; Tap feed
Tap
Cartridge kit for E-Pure 4-holder; Tap feed
Cartridge Kit