Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits.
Brand:  Thermo Scientific™ D5022
Code : NEW
Additional Details : Weight : 3.62870kg
 View more versions of this product
Product Code. 12341278
  • £747.76 / Each
Estimated Shipment: 28-10-2024
to see stock.
Add to basket
Description
Description
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Replace worn, outdated and otherwise ineffective cartridges with Thermo Scientific™ Barnstead™ E-Pure™ Cartridge Kits. These cartridge kits are classified based on type of feed water used; choose the appropriate kit according to your feed water and need for organic free water.
Specifications
Specifications
Cartridge kit for E-Pure 3-holder, Tap feed, Organic
Tap
Cartridge kit for E-Pure 3-holder, Tap feed, Organic free
Cartridge Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Services