missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems

Produce Type 1 ultrapure water with low operating costs using the Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification system.

Brand:  Thermo Scientific™ D4642-33

3997.00 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 11466348

  • £5710.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.

AquaPro™ digital on-line analytical measurement platform featuring:

  • Large color graphic display
  • Temperature readout
  • Temperature compensated
  • Calibratable

Features:
  • System achieves resistivities up to 18.2 MΩ-cm and TOC levels below 10ppb.
  • Virgin polypropylene water pathways prevent recontamination.
  • Recirculation pump continuously recirculates water throughout the entire system to maintain water purity and reduce the quantity of wastewater required for rinse-up.
  • System is wall-mountable and completely assembled for easy installation.
  • Quarter-turn quick release canisters allow for easy cartridge replacement.
  • 0.2μm absolute final filter removes bacteria and particulates that could alter your results.
  • Digital resistivity meter is automatically temperature-compensated to 25°C, preventing fluctuating readings due to temperature changes.

Ordering Information:

Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.

Includes:

Wall bracket

Requires:

O-rings included with cartridge pack (sold separately)

Recommended for:

AA, GS, buffer preps, feedwater for analytical instruments or labware washers

Note:

If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE, CSA
<1 CFU/mL
Tap or Pretreated
Type 1
18.2 Mohm.cm at 25°C
<10 ppb
100 psig (6.9 bar)
<1 μM/mL
7.5 in.
37.5 in.
28 in.
240 V
0.055μS/cm
No
Up to 2.5 L/min.
Optional
No
Water Purification System
4
7.5 x 37.5 x 28 in. (19 x 95.3 x 71.1 cm)
19 cm
95.3 cm
71.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers