Learn More
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
£67.72 - £980.96
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Description | Price | Quantity & Availability | ||||
10422962
|
Thermo Scientific™
D502124 |
Easypure™ II UV and Easypure II UV/UF | Cartridge pack; deionized feed |
£456.92
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||||
10895960
|
Thermo Scientific™
D50237 |
Easypure | Mixed Bed COOL Cartridge, 6 in, Ultrapure |
£67.72
Each |
In Stock
Log in to see stock. |
||||
10728154
|
Thermo Scientific™
D502125 |
Easypure™ II UV and Easypure II UV/UF | Cartridge pack; RO or distilled feed |
£456.92
Each |
Estimated Shipment: 09-01-2025
Log in to see stock. |
||||
10041672
|
Thermo Scientific™
D502126 |
Easypure™ II and Easypure UF | Cartridge pack; deionized feed |
£444.01
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
10133502
|
Thermo Scientific™
D502127 |
Easypure™ II and Easypure UF | Cartridge pack; RO or distilled feed |
£419.92
Each |
Estimated Shipment: 06-03-2025
Log in to see stock. |
||||
10498453
|
Thermo Scientific™
FL1192X1 |
Easypure™ UF and Easypure II UV/UF | Ultrafilter |
£980.96
Each |
In Stock
Log in to see stock. |
||||
11426348
|
Thermo Scientific™
D50230 |
Easypure II System, 2/3 Macroreticular Resin and 1/3 Granular, Activated Carbon, Organic/Colloidal Removal | Organic/Colloidal removal |
£139.40
Each |
In Stock
Log in to see stock. |
||||
10502074
|
Thermo Scientific™
D50245 |
EasyPure II RF, RF/UV system, Sanitation Cartridge | Sanitation |
£84.84
Each |
In Stock
Log in to see stock. |
||||
Description
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
- Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
- Type 1 filter
D502124:
- Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge
D502127:
- Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
- Type 1 filter
D502125:
- Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
- Ultralow filter with Organic Free Cartridge