Learn More
Thermo Scientific™ Barnstead™ Easypure™ II
This product has been discontinued. For additional information or alternate products please contact your sales representative.
Brand:  Thermo Scientific™ D7035
 View more versions of this product
Product Code. 10284992
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
This product has been discontinued. For additional information or alternate products please contact your sales representative. The recommended replacement for this product is Thermo Scientific™ Barnstead™ MicroPure™.
Features:
  • Each system produces ultrapure water that exceeds the quantitative specifications of Type 1 water as described in ASTM™ D1193, ISO 3696, and CLSI-CLRW.
  • Three cartridges in series provide increased capacity over similar systems.
  • A 0.2μm final filter at the point of use provides water free of particles and bacteria.
  • Quiet pump provides optimal flowrates and silent operation.
  • Standby mode recirculates purified water during inactivity to ensure purity and allows for on-demand use.
  • All components are constructed of inert materials, ensuring maximum ionic and organic purity.
  • Supplied wall bracket allows increased flexibility when choosing a location.
  • Integral low water and low pressure pump protectors protect the pump in the event feedwater is interrupted to the system.
Easypure II:
  • Base unit
  • For buffer prep, titration, solutions, AA
  • TOC 5 to 10ppb
Easypure II UV:
  • Ultralow organics
  • For HPLC, TOC, GC/MS
  • TOC 1 to 5ppb
Easypure II UFI:
  • Eliminates pyrogens
  • For cell and tissue culture, monoclonal antibody production, electrophoresis
  • TOC 5 to 10ppb
Easypure II UV:
  • Lowest levels of organics, pyrogens and nucleases
  • For PCR 2-D electrophoresis, cell culture, blotting
  • TOC 1 to 5ppb
Ordering Information:
Optional accessories include Remote Dispenser (0903515) and replacement consumables (UV lamps, ultrafilters and cleaning syringe); order separately. Contact your Sales Representative for details.
Includes:
Wall mounting bracket, cleaning syringe, UV lamp (Easypure II UV and UV/UF only), ultrafilter (Easypure II UF and UV/UF only).
Requires:
  • Cartridges and filters
Recommended for:
  • HPLC
  • ICP
  • GC/MS
  • Cell and tissue culture
  • PCR
Note:
  • Feedwater must be pre-treated by reverse osmosis, distillation or deionization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE, CSA
Discontinued; replacement product is 50132374
No
Up to 1 L/min.
18.2 Mohm.cm at 25°C
0 to 10 ppb
100 psig (6.9 bar)
RF/UV model
100/240 V 50/60 Hz
50/60 Hz
0.055 µS/cm
<1 CFU/mL
Pretreated
Type 1
No
Water Purification System
4°C to 40°C
<1 μM/mL
100/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products