Learn More
Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application.
Brand: Thermo Scientific™ D7034
Code : 03
Additional Details : Weight : 0.25000kg
Description
Customize your water purification system with the Thermo Scientific™ Barnstead™ Easypure™ RoDi Accessories and Consumables that best suit your application. Cartridge kit includes pre-filter, ultrapure mixed bed cartridge and Easypure high purity/low TOC cartridge. Other consumables include 0.2 micron final filter, ultraviolet lamp, Thermo Scientific™ Ventgard™ reservoir cap, reverse osmosis membrane, and set of three empty cartridges for cleaning.
Specifications
Empty Cartridges | |
Barnstead Easypure RoDi | |
Cartridge, Empty |