Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Brand:  Thermo Scientific™ D50281
Code : 03
Additional Details : Weight : 5.40000kg
 View more versions of this product
Product Code. 10024771
  • £903.51 / Each
In Stock
to see stock.
Add to basket
Description
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
Specifications
Deionized water feed ; Ultralow organics, type I water
Nanopure Water Purifications System
Deionized Water Feed