Learn More
Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Brand: Thermo Scientific™ D50282
Code : 03
Additional Details : Weight : 0.90000kg
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
- Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
R/O and distilled feed; Low organics, type I water | |
Nanopure Water Purifications System | |
R/O and Distilled Feed |