Learn More
Gilac™ Barrel with Tab Lid
Barrel with Tab Lid is a food contact certified plastic barrel, suitable for a large number of uses (brining, pickling etc.) when transporting and storing liquid or solid food products.
Brand: Gilac™ G032208
Additional Details : Weight : 1.96000kg
Description
- Food contact certified plastic barrel.
- Suitable for a large number of uses (brining, pickling etc.) when transporting and storing liquid or solid food products.
- Global and specific migration test approved.
- Contents are protected from contamination thanks to its watertight lid.
- Handling is easy and can be opened completely.
Specifications
Barrel | |
50 L | |
Alimentary or dangerous liquid |
13.2 gal. | |
White | |
HDPE |