Learn More
Corning™ Base Head Clamp
GREENER_CHOICE
For Use With: Holding Rod
Brand:  Corning™ 440140
Product Code. 10561758
  • £53.80 / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Corning offers a 2 diameter support rods. It is designed to be used with the Corning Temperature probe support clamp (Cat. No. 440141).
Note: Bosshead clamps to support rod; holding rod connects to bosshead. Temperature controller slides into holding rod.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Holding Rod
Clamp