Learn More
Daniels Healthcare™ Mini Bin Sharps Container Base
Daniels Healthcare™ Mini Bin Sharps Container Base is designed to meet demands for strength, simplicity, and value.
Brand:  Daniels Healthcare™ DD498
Product Code. 12913515
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Designed to meet demands for strength, simplicity and value.
  • Containers offer outstanding protection against penetrating sharps and leaking fluids due to the rigid polypropylene construction and wall thickness.
  • Containers are manufactured to BS 7320:1990, ISO 9002 standards and UN Packing Group II approved.
  • Containers have all British Standard markings, user instructions and a visible fill line
Specifications
Specifications
Safe disposal of sharps
Mini Sharps Bin / Bin Top