missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcold™ Basic Freezers
Providing a large capacity combined with good economical value for laboratory and general use. Labcold™ Basic Freezers are suitable for the storage of certain temperature-sensitive materials such as enzymes.
£635.00 - £3176.15
Specifications
Type | General Purpose Freezer |
---|---|
Temperature Range | -18°C to -25°C |
Door Style | Solid |
Frequency | 50/60 Hz |
Product Code | Brand | Capacity (Metric) | Dimensions (D x W x H) Interior | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (D x W x H) Interior | Voltage | Price | Quantity & Availability | |||||
11548944
|
Labcold™
RLVF1825 |
505 L | 545 x 645 x 1490 mm | 230 V | N/A | ||||||
12328067
|
Labcold™
RLVL03203 |
70 L | 365 x 305 x 370 mm | 240 V | N/A | ||||||
Description
- Door lock
- CFC and HCFC free
- Temperature range of -18° to -25°C
- Analog temperature display
- High temperature warning light
- Operator-controlled defrost
Specifications
General Purpose Freezer | |
-18°C to -25°C | |
Solid | |
50/60 Hz |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labcold™ Basic Freezers