Learn More
Memmert™ Basic Series Water Baths
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
Brand:  Memmert™ WNB7
 View more versions of this product
Product Code. 10738303
Description
Description
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Specifications
Microprocessor PID
CE, VDE, GS
For research laboratories, food and beverage, temperature control
WNB 7
±0.1°C
240 mm
210 mm
140 mm
14 kg
230 V
50/60 Hz
7 L
Digital LED
Stainless Steel
95°C
468 mm
356 mm
238 mm
240 mm
11 kg
1.2 kW
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ Basic Series Water Baths > Capacity: 7L