Learn More
Thermo Scientific™ Basic Vortex Mixers
The Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.
Brand: Thermo Scientific™ 88882012
237.60 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Includes: Standard package includes the Rubber platform 88882120 x 1
Description
The Thermo Scientific™ Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.
- Digital Speed display
- Continuous and Touch modes
- Speed range: 200 to 3000rpm
- Speed Accuracy: ±5% rpm
- Convenient Speed adjustments by knob control
- Stable for long-term operations
- Multiple accessories for different vessels
Order Info
Please order various accessories as the adapter for multiple applications
Specifications
4 mm | |
200 to 3000 rpm | |
Tubes | |
EU, UK, CN Plug | |
ABS housing | |
11.4 in. | |
14.99 lb. | |
14.33 lb. | |
Basic Vortex Mixer | |
Accessories: 88882120, 88882121, 88882122, 88882123, 88882124, 88882125, 88882126, 88882127 | |
IP 32 | |
19 | |
2 Years |
0.5 kg | |
100/240 V | |
CE | |
50/60 Hz | |
290 x 270 x 240 mm (11.4 x 10.7 x 9.4 in.) | |
290 mm | |
6.8 kg | |
6.5 kg | |
Digital | |
Rubber platform 88882120 | |
DC motor | |
Touch, Continous |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.