missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ Batch Sampling Oven Thermometer with Calibration Certificate
Precision test oven thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with traceable calibration.
£145.00 - £186.00
Specifications
Bottle Fill | Aluminium oxide crystals |
---|---|
Bottle Size | 30 mL |
For Use With (Application) | Oven Batch Sampling |
Height (English) | 5.71 in. |
Height (Metric) | 14.5 cm |
Product Code | Brand | Calibrated | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Calibrated | Price | Quantity & Availability | |||||
17873288
|
Brannan™
17/102/2 + 49/503/0 |
3-point UKAS |
£186.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17813058
|
Brannan™
17/102/2 + 49/101/0 |
3 Point Traceable |
£145.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Non-hazardous LO-tox™ filling
- Temperature measurement unaffected by short term fluctuations
- Temperature range: +20°C to +130°C
- Supplied with calibration and statement of accuracy
- Made in UK
Specifications
Aluminium oxide crystals | |
Oven Batch Sampling | |
14.5 cm | |
14.5 cm | |
Aluminium oxide crystals | |
91 g | |
266°F | |
68°F | |
68°F to 266°F | |
None | |
1.0°C | |
3.5 cm |
30 mL | |
5.71 in. | |
Polypropylene bottle and glass thermometer | |
Glass | |
3.2099 oz. | |
0.14 in. | |
+130°C | |
+20°C | |
+20°C to +130°C | |
± 2°C | |
None |