missing translation for 'onlineSavingsMsg'
Learn More

Brannan Batch Sampling Thermometer, Incubator

Product Code. 16504822
Click to view available options
Temperature Range (Metric):
18°C to 50°C
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16504822

Brand: Brannan 22/105/2

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These precision test thermometers are sealed into a polypropylene bottle and are ideally suited to monitor the temperature of samples in controlled storage

  • Non hazardous LO-toxTM filling
  • Temperature measurement unaffected by short term fluctuations
  • Supplied with statement of accuracy
  • Bottle should be filled with monopropylene glycol or other heat transfer medium before use
  • Made in UK

Specifications

For Use With (Application) Measures Temperature of Samples in Controlled Storage
Includes Polypropylene Bottle
Length (Metric) 15.3 cm
Material Glass
Thermometer Fill Lo-Tox Filling
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 64.4°F
Min. Temperature (Metric) 18°C
Temperature Range (English) 64.4°F to 122°F
Temperature Range (Metric) 18°C to 50°C
Calibrated No
For Use With (Equipment) Incubator
Accuracy ±0.5°C
Subdivision 1.0°C
Display Analog
Width (Metric) 3.7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.