Learn More
Fisherbrand™ Battery Acid Specialty Spill Kit
Spill kit designed to pick up battery acid on both smooth and rough surfaces while remaining chemically inert.
Brand:  Fisherbrand™ FBSKHBATTE
Product Code. 16563852
  • £220.00 / 1 set
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Place near a battery swap station and around your facility
  • Allows you to be prepared should a spill occur
  • Designed to pick up battery acid on both smooth and rough surfaces
  • Chemically inert
Specifications
Specifications
Chemical Sorbent
1 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper, 1 x SF-JAR-CS (149265)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : sodium carbonate