Learn More
BAX Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 50599-2-IG-150UL
322.05 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
The BAX antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BAX. This antibody recognizes human, mouse, rat antigen. The BAX antibody has been validated for the following applications: FC, IP, WB, ELISA analysis.
Specifications
BAX | |
Polyclonal | |
Unconjugated | |
150 μL | |
581 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Flow Cytometry, ELISA | |
340 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |