Learn More
Bollé Safety BAXTER Foam and Strap Kit Product Code.: 16610413
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
Bollé Safety BAXTER Foam and Strap Kit
Product Code. 16610413
Product Code. 16610413
Brand: Bollé Safety BAXKITFS
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
Add to basket
BAXTER, the alternative to the TRACKER model, offers total comfort thanks to its highly wraparound frame. With its waterproof foam and ingenious system for attaching the strap, BAXTER offers effective protection from sprays of solids and liquids, and from dust. BAXTER will soon be available in the RX range.
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Platinum
Color Black
Product Type Spare parts
Disposable Reusable
Frame Material Polycarbonate
Product Content Correction
Product Title
Bollé Safety BAXTER Foam and Strap Kit >