Learn More
RETSCH BB 100 Model Jaw Crushers
Designed for the rapid, effective crushing and pre-crushing of medium-hard, hard, brittle and tough materials. Retsch™ BB 100 Model Jaw Crushers are ideal for sample preparation in laboratories and industrial plants.
Brand:  RETSCH 20.052.0009
 View more versions of this product
Product Code. 10486611
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Excellent crushing performance
  • Wide range of materials for contamination free grinding
  • Wear compensation with zero-point adjustment
  • Continuous gap width setting
  • Overload protection
  • No-rebound feed hopper with quick-release clamp
  • Brake motor with safety switch
  • Easy-to-clean crushing chamber
  • Continuous grinding
  • Connector for dust extraction
Specifications
Specifications
Jaw Crusher
Analog
For pre-crushing hard, brittle products with a degree of hardness >3 on the Mohs' Scale
50 Hz
800 mm
BB 100
230 V
320 mm
800 x 320 x 960 mm
230 V, 50 Hz
960 mm
IP54
Tungsten Carbide
1-phase motor / 3-phase motor
750 W
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RETSCH BB 100 Model Jaw Crushers > Material: Tungsten Carbide/Tungsten Carbide; Electrical Requirements: 230V, 50Hz