missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Crystal™ ID Systems Accessories
Click to view available options
Product Type:
Anaerobic Nh inoculum
Calibration Standard
Microbiology Media
Quantity:
10 Pack
Each
Unit Size:
Each
Pack of 10
Description
BBL Crystal MIND Software
- Interprets the test results and provides specimen identification
- Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
- For facilitated viewing and recording of chromogenic and fluorogenic reactions
- Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard
†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.
Specifications
Specifications
| Form | Liquid |
| Product Type | Microbiology Media |
| For Use With (Equipment) | Crystal™ ID system |
| Quantity | 10 Pack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction