Learn More
BD BBL™ Dehydrated Culture Media: Soy Agar with Lecithin and Polysorbate 80
Dehydrated media for isolation, detection and cultivation of wide variety of microorganisms
Brand: BD 212263
Additional Details : CAS Number : 8013-01-2 Weight : 0.01000kg
Description
Detection of microorganisms on surfaces sanitized with quaternary ammonium compounds
Certifications
CE, CCAM
Specifications
CCAM | |
Soy Agar with Lecithin and Polysorbate 80 | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/mL, Polysorbate 80 5g/L, Agar 15g/L | |
Pail |
Dehydrated | |
Agar | |
Detection and enumeration of microorganisms present on surfaces of sanitary importance | |
5 lb. |