missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline
Fluorescent Antibody reagents
£50.15 - £118.91
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Content And Storage | <30°C |
Additive | Phosphate Buffered Saline |
Form | Dehydrated |
For Use With (Application) | Fluorescent Antibody reagents |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11742774
|
BD
223142 |
Bottle | 100 g |
£50.15
100g |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
11752774
|
BD
223143 |
Vial | 10 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
ISO/DIS, 11014 | |
Phosphate Buffered Saline | |
Fluorescent Antibody reagents |
<30°C | |
Dehydrated |