missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Gram Stain Kits

Product Code. 11748213
Click to view available options
Type:
Gram Stain Kit (Stabilized)
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11748213

Brand: BD 212539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to stain microorganisms from cultures or specimens by the differential Gram method

  • Complete stain kits reduce the mess associated with Gram staining procedures
  • Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
  • Stabilized Iodine Solution requires no mixing and has a long shelf life

TRUSTED_SUSTAINABILITY

Specifications

Type Gram Stain Kit (Stabilized)
Quantity 1 Kit
Certifications/Compliance CE
Includes 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin
For Use With (Application) Used to stain microorganisms from cultures or specimens by the differential gram method
Stain Type Gram Stain
compliance-icons compliance-icons
Product Identifier
  • Gram Stain Set w/Stabilized Iodine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : isopropanol, acetone, ethanol, methanol,isopropanol, ethanol, methanol, C.I. Basic Violet 3, Polyvi
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.