missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ MycoPrep™ Mycobacterial System Digestion/Decontamination Kits

No weighing and no messy mixing

Brand:  BD 240862


 View more versions of this product

Product Code. 15324564

  • £178.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Provide one of the most widely used, CDC-recommended formulations for effective mycobacterial specimen processing
  • Convenient, all-inclusive kits are an alternative to traditional methods of preparing specimen digestion/decontamination reagents
  • Complete with bottles of NALC-NaOH Solution and pre-measured packets of Phosphate Buffer (pH 6.8)
  • Ensure standardization by eliminating the need to weigh, reconstitute or inventory separate ingredients
  • Remain stable for up to 24 hours after activation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Specimen Collection Kit
For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis
15°C to 25°C
CE
10 x 75 mL bottles of NALC-NaOH solution and 5 packets Phosphate Buffer (ph 6.8)
75 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • kit mycoprep

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers