missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Lowenstein-Jensen Media

Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species

£392.00

Specifications

Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Lowenstein-Jensen Media
Formulation Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 30g/L, Malachitegreen 0.4g/L, Glycerol 12mL, Whole Egg 1000mL
View More Specs

Products 1
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
11938536
View Documents Promotion Details
BD
220909
Slants (A tubes) 100/Pk.
£392.00
Pack of 100
View alternative products
Save Up to 
»
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
 
Description

Description

  • Inspissated, egg-based media
  • Available as Mycoflask™ bottles, prepared slants and bottles

Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.

Specifications

Specifications

2°C to 8°C
United States
Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 30g/L, Malachitegreen 0.4g/L, Glycerol 12mL, Whole Egg 1000mL
Liquid
Lowenstein-Jensen Media
Cultivation of Mycobacterium tuberculosis and other mycobacterial species
Videos
SDS
Documents

Documents

Product Certifications

For in vitro diagnostic use.