missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Prepared Plated Chromogenic Media: MI Agar

Culture media tests drinking water for the presence of E.coli and total coliforms with membrane filtration method

Brand:  BD 214986

 View more versions of this product

Product Code. 17343650

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Detects degree of water quality deterioration in drinking distribution system enumerates, and monitors the progress in remediation. Provides precise and quantifiable results; also less time consuming and costly than other techniques.

  • Chromogenic-fluorogenic medium for detecting total coliforms and Escherichia coli in 24 hours or less
  • Under ambient light, E. coli exhibits bluish-gray appearance; under long-wave UV light, it displays blue-green fluorescence
  • Coliform are cream under ambient light, and display blue-white fluorescence under long-wave UV light
  • Conforms to EPA Method 1604 for monitoring drinking water under the Total Coliform Rule, and source water under the Surface Water Treatment

Applications:

For testing ambient and potable water, treatment plant effluents, and beverage and pharmaceutical manufacturing

Order Info

Available dehydrated or in prepared 15 x 60mm plates.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

US, EPA
25°C
United States
No. 3 Proteose Peptone 5g/L, Yeast extract 3g/L, Lactose 1g/L, 4-Methylumbelliferyl-beta-D-Galactopyranoside 0.1g/L, Indoxyl-beta-D-Glucuronide 0.32g/L, Sodium Chloride 7.5g/L, Dipotassium Phosphate 3.3g/L, Monopotassium Phosphate 1g/L, Sodium Lauryl Sulfate 0.2g/L, Sodium Desoxycholate 0.1g/L, Agar 15g/L, Cefsulodin 5mg/L
Plate
Beige
Prepared Plate
MI Agar
To detect and enumerate Escherichia coli and total Coliforms in drinking Water by the membrane filtration technique
20/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BD BBL™ Prepared Plated Chromogenic Media: MI Agar > 20/Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.