Learn More
Brady™ BBP37 Multicolor and Cut Sign and Label Printer for UK
The BBP37 Label Printer offers outstanding color and cutting capabilities with automatic label setup and intuitive touchscreen, enabling you to make signs on demand in any color and shape.
Brand: Brady™ 145995
Code : NEW
Additional Details : Weight : 21.40000kg
Description
The BBP™37 gives you the versatility to print all your basic label types from one machine, saving costs, reducing downtime and simplifying your labeling.
- Over 800 varieties of labels, including external-feed bulk rolls and custom parts for general identification, wire marking, asset tracking and more
- Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
- Centering ribbon spindle virtually eliminates ribbon wrinkle
- Compact size won't clutter your workplace or bench
- Comes with QWERTY keyboard and free Brady™ Workstation Basic Design Software Suite
Specifications
Color Touchscreen | |
19 in. | |
11 in. | |
15 in. | |
Not Autoclavable | |
BBP37 Printer, UK Power Cord, Brady™ Workstation Basic Suite, USB Cable, Cleaning Kit, Cutter Cleaning Tool, Quick Start Guide | |
512 MB | |
127 mm/s | |
Bluetooth™, Ethernet, USB | |
36 lb. | |
Cold Temperature Labeling, Electrical Labeling, Hazard and Safety Identification | |
110/240 V |
Thermal Transfer | |
483 mm | |
279.4 mm | |
363 mm | |
Multi-color | |
Bulgarian, Hungarian, Italian, Danish, Dutch, Portuguese, Estonian, Finnish, Slovakian, Slovenian, Spanish, Croatian, Swedish, Turkish, Czech, Danish, Dutch, English, Estonian, Finnish, French, German | |
BBP37 | |
300 dpi | |
10°C to 40°C | |
16.3 kg | |
1 Year |