missing translation for 'onlineSavingsMsg'
Learn More

BC 11 hydrobromide, Tocris Bioscience™

Product Code. 18034152
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective urokinase (uPA) inhibitor

Selective urokinase (uPA) inhibitor (IC50 = 8.2 μM). Exhibits no activity at 8 other related enzymes. Inhibits clot lysis with no effect on clot formation. Also decreases viability of MDA-MB231 breast cancer cells in vitro.

Chemical Identifiers

CAS 443776-49-6
Molecular Formula C8H12BBrN2O2S
Molecular Weight (g/mol) 290.97
InChI Key PAFZAMOVHIRQOD-UHFFFAOYSA-N
Synonym bc 11 hydrobromide, carbamimidothioic acid 4-boronophenyl methyl ester hydrobromide
PubChem CID 78243712
IUPAC Name [4-(carbamimidoylsulfanylmethyl)phenyl]boronic acid;hydrobromide
SMILES B(C1=CC=C(C=C1)CSC(=N)N)(O)O.Br

Specifications

Quantity 10 mg
Formula Weight 290.97
Percent Purity >98%
Chemical Name or Material BC 11 hydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'