missing translation for 'onlineSavingsMsg'
Learn More

Bcl-2 Antibody (BCL2/2210R), Novus Biologicals™

Rabbit Monoclonal Antibody

£236.00 - £423.00

Specifications

Antigen Bcl-2
Clone BCL2/2210R
Concentration 0.2 mg/mL
Dilution Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655416
View Documents
Novus Biologicals
NBP2-75761-20ug
20 μg
£236.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18638188
View Documents
Novus Biologicals
NBP2-75761-100ug
100 μg
£423.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Bcl-2
0.2 mg/mL
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Unconjugated
Rabbit
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Oncogenes, Tumor Suppressors
10 mM PBS with 0.05% BSA
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Recombinant full-length human bcl-2 protein (Uniprot: P10415)
Primary
Store at 4°C.
BCL2/2210R
Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Human
P10415, P10415, P10415, P10415
596
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.