Learn More
Abnova™ BCL2 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FS0052.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16013495
  • £2030.00 / 200µL
Estimated Shipment: 27-08-2024
to see stock.
Specifications
Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
Bcl-2
596
200 μL
Tube
B-cell CLL/lymphoma 2
BCL2
DNA/RNA