Learn More
Berkshire™ BCR™ Cleanroom MyNotes
BIOPHARMA
Our BCR™ Mynotes are made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release
Brand:  Berkshire™ MNBKA4CR
398.05 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 17336449
  • £435.00 / Pack of 20
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
They are practical for note taking and data recording in the cleanroom. Bond is made with the purest quality cellulose from specially selected sustainable forests and is coated with a proprietary formula to minimise particle and fibre release.
Specifications
Specifications
Cellulose
ISO Class 3 - 7, Cleanroom Class 1 - 10,000, EU Grade B/C/D
A4
College Ruled
50 Sheets/book